Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Lipomed d-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
Lipomed d-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
Lipomed d-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
77976-095
:
LGC Standards
New Product
:
Lipomed d-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
77976-095
:
LGC Standards
:
LPM-AMP-96-HC-1LM
New Product
:
Matrix:
Methanol
CAS No.:
1462-73-3
Description:
d-Amphetamine.HCl, 1 mg/ml in Methanol (as free base)
Size:
1 ml