$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC N,N'-Dicyclohexylurea
TRC N,N'-Dicyclohexylurea
77982-487
:
LGC Standards
New Product
:
TRC N,N'-Dicyclohexylurea
77982-487
:
LGC Standards
:
TRC-D439365-25G
New Product
:
CAS No.:
2387-23-7
Description:
N,N'-Dicyclohexylurea
Size:
25 g