Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lipomed Meperidine-D4.HCl (Pethidine-D4.HCl), 1 mg/ml in Methanol (as free base)
Catalog # 77976-203
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lipomed Meperidine-D4.HCl (Pethidine-D4.HCl), 1 mg/ml in Methanol (as free base)
Catalog # 77976-203
Supplier:  LGC Standards
Supplier Number:  LPM-MEP-1536-HC-1LM
New Product
CAS Number:  

Specifications

  • Matrix:
    Methanol
  • CAS No.:
    2748469-74-9
  • Description:
    Meperidine-D4.HCl (Pethidine-D4.HCl), 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml

Specifications

About this item