Order Entry
Puerto Rico
ContactUsLinkComponent
Iron Total Ferrous Instrumental Test
Iron Total Ferrous Instrumental Test
Catalog # 77674-808
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Iron Total Ferrous Instrumental Test
Catalog # 77674-808
Supplier:  CHEMetrics
Supplier Number:  K-6203
New Product
CAS Number:  

Specifications

  • Chemical method:
    Phenanthroline (total and ferrous)
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Surface and ground water;Sea water;Drinking water

Specifications

About this item

  • Range: 0 to 6.00 ppm Method: Phenanthroline (total and ferrous)
Delivery: Kit K-6203 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, activator solution, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.