Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Dechloro-3-bromo Trazodone Hydrochloride
TRC 3-Dechloro-3-bromo Trazodone Hydrochloride
77981-204
:
LGC Standards
New Product
:
TRC 3-Dechloro-3-bromo Trazodone Hydrochloride
77981-204
:
LGC Standards
:
TRC-D226260-50MG
New Product
:
CAS No.:
1263278-80-3
Description:
3-Dechloro-3-bromo Trazodone Hydrochloride
Size:
50 mg