$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Rivastigmine N-Oxide (Technical Grade)
TRC Rivastigmine N-Oxide (Technical Grade)
Catalog # 77992-846
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Rivastigmine N-Oxide (Technical Grade)
Catalog # 77992-846
Supplier:
LGC Standards
Supplier Number:
TRC-R540900-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
1369779-37-2
Description:
Rivastigmine N-Oxide (Technical Grade)
Size:
2.5 mg
Specifications
About this item
Product Details
Product Details