Paragon Scientific Relative Density 20 °C (Nominal: 0.7769 @ 20 °C)
Catalog # 77975-952
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Relative density 20 °C (Nominal: 0.7769 @ 20 °C), Standard type: Single component
- Size:500 ml
Specifications
About this item
- Shipping conditions: Room temperature