$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Clotrimazole
TRC Clotrimazole
:
LGC Standards
New Product
TRC Clotrimazole
:
LGC Standards
New Product
CAS No.:
23593-75-1