Lipomed Dihydrocodeine.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-107
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:36418-29-8
- Description:Dihydrocodeine.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml