Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Butanilicaine Hydrochloride
TRC Butanilicaine Hydrochloride
77979-166
:
LGC Standards
New Product
:
TRC Butanilicaine Hydrochloride
77979-166
:
LGC Standards
:
TRC-B693055-10MG
New Product
:
CAS No.:
6027-28-7
Description:
Butanilicaine Hydrochloride
Size:
10 mg