TRC Metamizol Sodium Monohydrate
New Product
- CAS No.:5907-38-0
Frequently Bought Together
pH Gradient Buffers
: Thermo Scientific
Accessories for Gas Clean Purification System, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Replacement Frits for Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Prep HPLC Columns, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Cleaning and Maintenance Supplies for LC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Cylinder Supplies, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Guard Cartridge Holder, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Column Thermostat Accessories for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
O-Rings and Fittings for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)