ContactUsLinkComponent
Total Alkalinity Titrets® Test Kit (50 to 500 ppm)
  77674-882
New Product
 :  
Total Alkalinity Titrets® Test Kit (50 to 500 ppm)
  77674-882
 :  K-9815
New Product
 :  
  • Chemical method:
    Acid titrant with pH indicator
  • Application:
    Drinking, surface and saline water;Domestic and industrial waste water;Boiler and cooling water
Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE
  • Range: 50 to 500 ppm as CaCO₃ (Total) MDL: 50 ppm Method: Acid titrant with pH indicator Increments: 50, 55, 60, 65, 70, 75, 80, 90, 100, 125, 150, 175, 200, 250, 350, 500 ppm
 : The K-9815 total alkalinity Titrets® Kit comes in a cardboard box and contains everything needed to perform 30 total alkalinity titration tests: thirty Titrets® ampoules with valve assemblies, activator solution, titrettor, 25 ml sample cup, and instructions. This test features the common calcium carbonate titration for measuring alkalinity packaged in a convenient Titrets® ampoule.