$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Avenanthramide C
TRC Avenanthramide C
Catalog # 77977-926
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Avenanthramide C
Catalog # 77977-926
Supplier:
LGC Standards
Supplier Number:
TRC-A794760-1MG
New Product
CAS Number:
Specifications
CAS No.:
116764-15-9
Description:
Avenanthramide C
Size:
1 mg
Specifications
About this item
Product Details
Product Details