$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed 20 mg/dl Aq. Ethanol Standard Solution, 20 mg/dl in Water
Lipomed 20 mg/dl Aq. Ethanol Standard Solution, 20 mg/dl in Water
77976-155
:
LGC Standards
New Product
:
Lipomed 20 mg/dl Aq. Ethanol Standard Solution, 20 mg/dl in Water
77976-155
:
LGC Standards
:
LPM-ETH-020-1ML-5AMP
New Product
:
Matrix:
Water
CAS No.:
64-17-5
Description:
20 mg/dL Aq. Ethanol Standard Solution, 20 mg/dL in Water
Size:
5×1 ml