Monochloramine Instrumental Test
Catalog # 77674-824
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Hydroxybenzyl Alcohol (HBA)
- Application:Drinking water;Clean surface water;Waste water;Storm water
Specifications
About this item
- Range: V-2000: 0 to 15.0 ppm, V-3000 and Spec: 0 to 8.00 ppm as NH₂Cl-Cl₂ Method: Hydroxybenzyl Alcohol (HBA)
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, stabilizer solution, catalyzer solution, 25 ml sample cup, ampoule blank, and instructions.