Paragon Scientific Type S600HT, High Temperature Viscosity (Ranges: 100 to 150 °C)
Catalog # 77975-999
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type S200HT, High Temperature Viscosity (Ranges: 100...150°C)
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component