TRC 1-Palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:2680-59-3
Specifications
Frequently Bought Together
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Upchurch Scientific® Coned Port Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Adapters and Unions for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Disintegration Tubes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Plugs and Caps, IDEX Health and Science
Supplier: Upchurch Scientific
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Prep HPLC Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX for NP/SFC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
pH Gradient Buffers
Supplier: Thermo Scientific
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Source Lamps for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Pre-Optics Cone and Windows for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)