CHEMets® Iron Total Soluble HR Visual Test Kit 250 to 2500 ppm
Catalog # 77674-804
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Phenanthroline (total and soluble)
- Application:Industrial waste water influent and effluent;Industrial process water;Surface and ground water;Sea water;Drinking water
Specifications
About this item
- Method: Phenanthroline (total and soluble) Range: 250 to 2500 ppm MDL: 250 ppm
Delivery: The K-6020B Iron (total and soluble) HR CHEMets® test kit (250 to 2500 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, pipettor tips, 100 µl fixed volume pipet, iron activator solution, and instructions.
Note: Kit contains only enough activator solution to perform approximately 7 total iron tests.