Lipomed Cocaine.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-128
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:53-21-4
- Description:Cocaine.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml