Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Cone and Plate (Nominal: 20000 mPas @ 25 °C), Viscosity
Paragon Scientific Cone and Plate (Nominal: 20000 mPas @ 25 °C), Viscosity
Catalog # 77975-479
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Cone and Plate (Nominal: 20000 mPas @ 25 °C), Viscosity
Catalog # 77975-479
Supplier:  LGC Standards
Supplier Number:  ALK-2162/20
New Product
CAS Number:  

Specifications

  • Description:
    Cone and plate (nominal: 20000 mPas @ 25 °C), viscosity
  • Size:
    500 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component