Chlorine Dioxide Comparator Round 0 to 2 ppm
77674-602
New Product
:
- Chemical method:DPD
- Cat. No.:77674-602
- Increments:0, 0.2, 0.4, 0.6, 0.8, 1.2, 1.6, 2.0 ppm
- Range:0 - 2.0 ppm
C-2702 Low Range Chlorine Dioxide Comparator is a component of K-2705 Chlorine Dioxide CHEMets Visual Test Kit and is designed to be compared against the color formed in R-2705 Chlorine Dioxide CHEMets Refill. The comparator's range is applicable for many disinfection and drinking water applications. Color standards are pink in color and cover a range of 0 to 2.0 ppm chlorine dixoide (ClO₂).