Paragon Scientific Type RT350, Rotational Viscosity
Catalog # 77976-036
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type RT350, rotational viscosity
- Size:600 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component