Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Phenols HR Visual Test Kit 0 to 300 ppm
CHEMets® Phenols HR Visual Test Kit 0 to 300 ppm
Catalog # 77674-856
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Phenols HR Visual Test Kit 0 to 300 ppm
Catalog # 77674-856
Supplier:  CHEMetrics
Supplier Number:  K-8020D
New Product
CAS Number:  

Specifications

  • Chemical method:
    4-Aminoantipyrine
  • Application:
    Domestic and industrial waste water;Surface water;Drinking water;Saline water;Disinfection control;Citrus fruit disinfection

Specifications

About this item

  • Method: 4-Aminoantipyrine Range: 0 to 300 ppm MDL: 25 ppm
Delivery: The K-8020D Phenols HR CHEMets® test kit (0 to 300 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, 1.0 ml syringe, and instructions.