ContactUsLinkComponent
Nitrate Comparator Flat 0 to 4.5 ppm
Nitrate Comparator Flat 0 to 4.5 ppm
Catalog # 77674-658
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Nitrate Comparator Flat 0 to 4.5 ppm
Catalog # 77674-658
Supplier:  CHEMetrics
Supplier Number:  C-6904
New Product
CAS Number:  
Specifications
  • Chemical method:
    Cadmium Reduction
  • Cat. No.:
    77674-658
  • Increments:
    0, 0.4, 0.7, 1.0, 1.4, 1.8, 2.5, 3.5, 4.5 ppm
  • Range:
    0 - 4.5 ppm
Specifications
About this item
C-6904 Nitrate Comparator is a component of K-6904 Nitrate CHEMets Visual Test Kit and is designed to be compared against the color formed in R-7002 Nitrate CHEMets Refill. The comparatorís range is applicable to many environmental applications. Color standards are pink to orange in color and cover a range of 0 to 4.5 mg/L nitrate (NO₃-).