ContactUsLinkComponent
Molybdate Comparator Flat 2 to 24 ppm
Molybdate Comparator Flat 2 to 24 ppm
Catalog # 77674-652
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Molybdate Comparator Flat 2 to 24 ppm
Catalog # 77674-652
Supplier:  CHEMetrics
Supplier Number:  C-6702
New Product
CAS Number:  
Specifications
  • Chemical method:
    Catechol
Specifications
About this item
C-6702 Molybdate Comparator is a component of K-6702 Molybdate CHEMets Visual Test Kit and is designed to be compared against the color formed in R-6702 Molybdate CHEMets Refill. The comparator's range is applicable to many industrial water treatment applications. Color standards are yellow-orange in color and cover a range of 2 to 24 mg/L molybdenum (Mo).