Lipomed Venlafaxine-D6.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-268
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1062606-12-5
- Description:Venlafaxine-D6.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml