TRC Yohimbine Hydrochloride
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:65-19-0
Specifications
Frequently Bought Together
CrossLab Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Headspace Sampler Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing and Fittings for 8453/8454 UV Systems
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
Intuvo Flow Chips, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Column Ferrules for Third Party GC Instruments
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab CE and CE/MS Filters and Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
O-Rings and Fittings for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)