$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 6-Methylene Progesterone Acetate
TRC 6-Methylene Progesterone Acetate
Catalog # 77988-961
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 6-Methylene Progesterone Acetate
Catalog # 77988-961
Supplier:
LGC Standards
Supplier Number:
TRC-M304220-10MG
New Product
CAS Number:
Specifications
CAS No.:
32634-95-0
Description:
6-Methylene Progesterone Acetate
Size:
10 mg
Specifications
About this item
Product Details
Product Details