TRC Tris(2-ethylhexyl) Phosphate-d51
New Product
- CAS No.:1259188-37-8
Frequently Bought Together
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Loops for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Purged Packed Inlet for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)