$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC p-Hydroxymephentermine Hydrochloride
TRC p-Hydroxymephentermine Hydrochloride
77986-167
:
LGC Standards
New Product
:
TRC p-Hydroxymephentermine Hydrochloride
77986-167
:
LGC Standards
:
TRC-H791720-1MG
New Product
:
CAS No.:
120583-18-8
Description:
p-Hydroxymephentermine Hydrochloride
Size:
1 mg