Lipomed Alcohol Mixture 4 (Acetone, Ethanol, Isopropanol, Methanol), 1 mg/ml in Water
Catalog # 77976-212
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Water
- Description:Alcohol Mixture 4 (Acetone, Ethanol, Isopropanol, Methanol), 1 mg/ml in Water
- Size:1 ml