$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed Tramadol.HCl, 1 mg/ml in Methanol (as free base)
Lipomed Tramadol.HCl, 1 mg/ml in Methanol (as free base)
77976-263
:
LGC Standards
New Product
:
Lipomed Tramadol.HCl, 1 mg/ml in Methanol (as free base)
77976-263
:
LGC Standards
:
LPM-TRA-779-HC-1LM
New Product
:
Matrix:
Methanol
CAS No.:
36282-47-0
Description:
Tramadol.HCl, 1 mg/ml in Methanol (as free base)
Size:
1 ml