Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Hydroxy Bromazepam 1.0 mg/mL in Methanol
TRC 3-Hydroxy Bromazepam 1.0 mg/mL in Methanol
TRC 3-Hydroxy Bromazepam 1.0 mg/mL in Methanol
Catalog # 77987-725
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-Hydroxy Bromazepam 1.0 mg/mL in Methanol
Catalog # 77987-725
Supplier:
LGC Standards
Supplier Number:
TRC-KIT0945-1X1ML
New Product
CAS Number:
Specifications
Matrix:
Methanol
CAS No.:
13132-73-5
Description:
3-Hydroxy Bromazepam 1.0 mg/ml in Methanol
Size:
1×1 ml
Specifications
About this item
Product Details
Product Details