Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Hexachlorophene
TRC Hexachlorophene
TRC Hexachlorophene
Catalog # 77985-899
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Hexachlorophene
Catalog # 77985-899
Supplier:
LGC Standards
Supplier Number:
TRC-H290810-5G
New Product
CAS Number:
Specifications
CAS No.:
70-30-4
Description:
Hexachlorophene
Size:
5 g
Specifications
About this item
Product Details
Product Details