Paragon Scientific Density 20 °C (Nominal: 1.1915 g/ml @ 20 °C)
77975-678
New Product
:
- Pack type:Glass bottle
- Description:Density 20 °C (Nominal: 1.1915 g/ml @ 20 °C), Standard type: Multi-component
- Size:500 ml
- Shipping conditions: Room temperature