Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 7-O-Demethyl Alpha-Narcotine
TRC 7-O-Demethyl Alpha-Narcotine
TRC 7-O-Demethyl Alpha-Narcotine
77981-431
:
LGC Standards
New Product
:
TRC 7-O-Demethyl Alpha-Narcotine
77981-431
:
LGC Standards
:
TRC-D231285-10MG
New Product
:
CAS No.:
68353-55-9
Description:
7-O-Demethyl Alpha-Narcotine
Size:
10 mg