Paragon Scientific IP 336, Sulfur set A in heavy mineral oil: S @ blank, 0.03, 0.05, 0.07, 0.1%
77975-801
New Product
:
- Matrix:Heavy Mineral Oil
- CAS No.:7704-34-9
- Description:IP 336, Sulfur set A in heavy mineral oil: S @ blank, 0.03, 0.05, 0.07, 0.1%
- Size:5×100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065