Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Poly(allylamine Hydrochloride) (Technical Grade)
TRC Poly(allylamine Hydrochloride) (Technical Grade)
77991-968
:
LGC Standards
New Product
:
TRC Poly(allylamine Hydrochloride) (Technical Grade)
77991-968
:
LGC Standards
:
TRC-P688303-250MG
New Product
:
CAS No.:
71550-12-4
Description:
Poly(allylamine Hydrochloride) (Technical Grade)
Size:
250 mg