$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Cis-Anethole
TRC Cis-Anethole
77977-598
:
LGC Standards
New Product
:
TRC Cis-Anethole
77977-598
:
LGC Standards
:
TRC-A653355-50MG
New Product
:
CAS No.:
25679-28-1
Description:
Cis-Anethole
Size:
50 mg