TRC 4-epi-Tetracycline Hydrochloride
New Product
- CAS No.:23313-80-6
Frequently Bought Together
CrossLab Nebulizers for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Torches for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Loops for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)