TRC (±)-Venlafaxine-d6 Hydrochloride (N,N-dimethyl-d6)
New Product
- CAS No.:1062606-12-5
Frequently Bought Together
Accessories for Stay Safe Caps for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Closures for 2 ml Crimp Top Vials, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Replacement Frits for Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
CrossLab Burners and Supplies for Agilent Instruments, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
CrossLab Fraction Collector Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Interface Cone Supplies for ICP-MS
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Heat Exchanger for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Tools, IDEX Health & Science
: Upchurch Scientific