$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt
TRC 5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt
77985-138
:
LGC Standards
New Product
:
TRC 5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt
77985-138
:
LGC Standards
:
TRC-F700980-10MG
New Product
:
CAS No.:
2469007-14-3
Description:
5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt
Size:
10 mg