Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Hydroxyoctanoic Acid-d12
TRC 3-Hydroxyoctanoic Acid-d12
TRC 3-Hydroxyoctanoic Acid-d12
77986-733
:
LGC Standards
New Product
:
TRC 3-Hydroxyoctanoic Acid-d12
77986-733
:
LGC Standards
:
TRC-H950807-1MG
New Product
:
CAS No.:
1215622-76-6
Description:
3-Hydroxyoctanoic Acid-d12
Size:
1 mg