Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Phosphate, Ortho HR Visual Test Kit (10 to 100 ppm)
CHEMets® Phosphate, Ortho HR Visual Test Kit (10 to 100 ppm)
Catalog # 77674-864
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Phosphate, Ortho HR Visual Test Kit (10 to 100 ppm)
Catalog # 77674-864
Supplier:  CHEMetrics
Supplier Number:  K-8520D
New Product
CAS Number:  

Specifications

  • Chemical method:
    Stannous chloride
  • Application:
    Domestic and industrial waste water;Industrial process water;Boiler water;Cooling water;Surface and ground water;Power generation;Pulp and paper industries;Food and beverages

Specifications

About this item

  • Method: Stannous Chloride Range: 10 to 100 ppm MDL: 10 ppm
Delivery: The K-8520D Phosphate, ortho HR CHEMets® test kit (10 to 100 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, 3.0 ml syringe, and instructions.