Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Hydrogen Peroxide HR Visual Test Kit 125 to 1250 ppm
CHEMets® Hydrogen Peroxide HR Visual Test Kit 125 to 1250 ppm
Catalog # 77674-790
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Hydrogen Peroxide HR Visual Test Kit 125 to 1250 ppm
Catalog # 77674-790
Supplier:  CHEMetrics
Supplier Number:  K-5520B
New Product
CAS Number:  

Specifications

  • Chemical method:
    Ferric thiocyanate
  • Application:
    Industrial effluent;Waste water;Sea water;Aseptic packaging;Food and beverages

Specifications

About this item

Delivery: The K-5520B hydrogen peroixde HR CHEMets® test kit (125 to 1250 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®;refill, flat comparator, 25 ml sample cup, 200 µl fixed volume pipet, and instructions.