Lipomed d,l-Amphetamine-D5.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-092
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:2714414-50-1
- Description:d,l-Amphetamine-D5.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml