Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Sodium Acetate
TRC Sodium Acetate
TRC Sodium Acetate
77993-220
:
LGC Standards
New Product
:
TRC Sodium Acetate
77993-220
:
LGC Standards
:
TRC-S610970-5G
New Product
:
CAS No.:
127-09-3
Description:
Sodium Acetate
Size:
5 g