Lipomed Duloxetine-D3.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-142
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1435727-97-1
- Description:Duloxetine-D3.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml