$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Diisobutyl Phosphate
TRC Diisobutyl Phosphate
77982-936
:
LGC Standards
New Product
:
TRC Diisobutyl Phosphate
77982-936
:
LGC Standards
:
TRC-D455480-50MG
New Product
:
CAS No.:
6303-30-6
Description:
Diisobutyl Phosphate
Size:
50 mg