$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4-tert-Butyl-d9-phenol-2,3,5,6-d4
TRC 4-tert-Butyl-d9-phenol-2,3,5,6-d4
77979-193
:
LGC Standards
New Product
:
TRC 4-tert-Butyl-d9-phenol-2,3,5,6-d4
77979-193
:
LGC Standards
:
TRC-B694106-10MG
New Product
:
CAS No.:
225386-58-3
Description:
4-tert-Butyl-d9-phenol-2,3,5,6-d4
Size:
10 mg